Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC683722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101008.1 | NP_001094478.1 | 5 | 425 | ||
XM_008758772.1 | XP_008756994.1 | 5 | 836 | |||
XM_008758773.1 | XP_008756995.1 | 5 | 833 | |||
XM_008758774.1 | XP_008756996.1 | 5 | 530 | |||
GenBank mRNA | BC098026.1 | - | 5 | 425 | 7455942 |