Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080936.1 | NP_001074405.1 | 2, 3 | 278 | ||
XM_006245063.2 | XP_006245125.1 | 8, 9 | 1309 | |||
XM_006245064.2 | XP_006245126.1 | 8, 9 | 1086 |