Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109449.2 | NP_001102919.1 | 5 | 953 | ||
XM_006243856.2 | XP_006243918.1 | 5 | 1228 | |||
XM_006243857.2 | XP_006243919.1 | 5 | 1011 | |||
GenBank mRNA | BC105621.1 | - | 5 | 953 | 7454021 |