Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128192.1 | NP_001121664.1 | 1 | 161 | ||
GenBank mRNA | BC167075.1 | - | 1 | 161 | 9093013 |