Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC684471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750810.3 | XP_003750858.3 | 2 | 1688 | ||
XM_008767723.1 | XP_008765945.1 | 2 | 2209 | |||
XM_008767724.1 | XP_008765946.1 | 3 | 1765 |