Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101013.2 | NP_001094483.2 | 8 | 1281 | ||
GenBank mRNA | BC107650.1 | - | 4 | 580 | ||
BC168141.1 | - | 8 | 1003 |