Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101013.2 | NP_001094483.2 | 10 | 1554 | ||
GenBank mRNA | BC107650.1 | - | 6 | 853 | ||
BC168141.1 | - | 10 | 1276 | |||
BG371460.1 | - | 1 | 400 |