Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172089.1 | NP_001165560.1 | 11, 12 | 1685 | ||
XM_006246396.2 | XP_006246458.1 | 11, 12 | 1694 | |||
XM_006246397.2 | XP_006246459.1 | 11, 12 | 1694 | |||
XM_006246398.2 | XP_006246460.1 | 11, 12 | 1691 | |||
XM_006246399.2 | XP_006246461.1 | 11, 12 | 1691 | |||
XM_006246400.2 | XP_006246462.1 | 11, 12 | 1691 | |||
XM_006246401.2 | XP_006246463.1 | 11, 12 | 1677 |