Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ss18l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750606.3 | XP_003750654.1 | 3 | 303 | ||
GenBank mRNA | FQ229074.1 | - | 3 | 249 |