Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110318.2 | NP_001103788.1 | 2 | 500 | ||
NM_001271085.1 | NP_001258014.1 | 2 | 500 | |||
GenBank mRNA | BC105914.1 | - | 2 | 361 | ||
BC160865.1 | - | 2 | 349 | 7324838 | ||
EU220430.1 | - | 2 | 500 |