Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748791.3 | XP_003748839.1 | 3 | 480 | ||
XM_006228514.2 | XP_006228576.1 | 3 | 1093 | |||
XM_006228517.2 | XP_006228579.1 | 4 | 650 | |||
XM_006228518.2 | XP_006228580.1 | 4 | 1890 | |||
XM_006228519.2 | XP_006228581.1 | 3 | 2819 | |||
XM_008759065.1 | XP_008757287.1 | 4 | 1733 |