Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750162.3 | XP_003750210.1 | 1 | 97 | ||
XM_008764676.1 | XP_008762898.1 | 1 | 97 | |||
XM_008764677.1 | XP_008762899.1 | 2 | 528 | |||
XM_008764678.1 | XP_008762900.1 | 1 | 97 |