Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750982.3 | XP_003751030.1 | 2, 3 | 1227 | ||
XM_003750983.3 | XP_003751031.1 | 3, 4 | 1293 |