Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168642.1 | NP_001162113.1 | 6 | 863 | ||
XM_006228106.2 | XP_006228168.1 | 6 | 906 | |||
XM_008758775.1 | XP_008756997.1 | 6 | 922 | |||
XM_008758776.1 | XP_008756998.1 | 4 | 547 |