Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082539.1 | NP_001076008.1 | 7 | 1235 | ||
NM_001082540.1 | NP_001076009.1 | 7 | 1145 | |||
NM_001082541.1 | NP_001076010.1 | 6 | 1088 | |||
NM_024404.2 | NP_077380.2 | 8 | 1292 | |||
GenBank mRNA | AB046615.1 | - | 8 | 1056 | ||
AB046616.1 | - | 7 | 950 | |||
AB046617.1 | - | 7 | 860 | |||
AB046618.1 | - | 6 | 803 | |||
FQ212501.1 | - | 5 | 545 |
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X16933.1 | - | 422 |