Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173103.1 | NP_775126.1 | 4 | 500 | ||
XM_006239320.2 | XP_006239382.1 | 7 | 3372 | |||
XM_008764293.1 | XP_008762515.1 | 6 | 3040 | |||
XM_008764294.1 | XP_008762516.1 | 6 | 1306 | |||
XM_008764295.1 | XP_008762517.1 | 5 | 1451 | |||
XM_008764296.1 | XP_008762518.1 | 5 | 1440 | |||
XM_008764297.1 | XP_008762519.1 | 4 | 1311 | |||
GenBank mRNA | BC081698.1 | - | 4 | 995 | ||
D26111.1 | - | 5 | 554 | |||
DQ437671.1 | - | 5 | 604 | |||
DQ437672.1 | - | 5 | 497 | |||
DQ437673.1 | - | 4 | 475 | |||
DQ437674.1 | - | 4 | 439 | |||
DQ437675.1 | - | 5 | 447 | |||
DQ437676.1 | - | 4 | 425 | |||
Z30663.1 | - | 4 | 500 |