Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024489.1 | NP_077815.1 | 7 | 2939 | ||
GenBank mRNA | AF091457.1 | - | 7 | 2939 |