Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030847.1 | NP_110474.1 | 2, 3 | 285 | ||
XM_006229158.2 | XP_006229220.1 | 2, 3 | 349 | |||
XM_008759418.1 | XP_008757640.1 | 2, 3 | 227 | |||
GenBank mRNA | BC088131.1 | - | 2, 3 | 281 | 7365466 | |
FQ233038.1 | - | 2, 3 | 272 | |||
Y10889.2 | - | 2, 3 | 285 |