Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035238.2 | NP_001030315.1 | 3 | 181 | ||
NM_031021.3 | NP_112283.1 | 3 | 232 | |||
XM_006256081.2 | XP_006256143.1 | 3 | 268 | |||
GenBank mRNA | BC078807.1 | - | 3 | 181 | 7133064 | |
CB743000.1 | - | 3 | 252 | |||
CB790784.1 | - | 3 | 278 | |||
EV778558.1 | - | 3 | 236 | |||
FQ214437.1 | - | 3 | 248 | |||
FQ224491.1 | - | 3 | 252 | |||
FQ230013.1 | - | 3 | 220 | |||
HE864425.1 | - | 2 | 116 | |||
HE864426.1 | - | 2 | 116 | |||
HE864431.1 | - | 116 | ||||
HE864432.1 | - | 116 | ||||
HE864433.1 | - | 116 | ||||
HE864434.1 | - | 116 |