Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031087.2 | NP_112349.2 | 3, 4 | 445 | ||
XM_006250405.2 | XP_006250467.1 | 5, 6 | 596 | |||
XM_006250407.2 | XP_006250469.1 | 4, 5 | 506 | |||
XM_006250408.2 | XP_006250470.1 | 3, 4 | 421 | |||
XM_006250410.2 | XP_006250472.1 | 4, 5 | 1189 | |||
XM_008769876.1 | XP_008768098.1 | 4, 5 | 1188 | |||
XM_008769877.1 | XP_008768099.1 | 5, 6 | 736 | |||
XM_008769878.1 | XP_008768100.1 | 3, 4 | 1814 | |||
GenBank mRNA | AB004454.1 | - | 2, 3 | 355 | ||
BC078805.1 | - | 3, 4 | 445 | 7109917 | ||
D83700.1 | - | 2, 3 | 348 | |||
FQ219614.1 | - | 5, 6 | 564 | |||
X99267.1 | - | 3, 4 | 468 |