Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031091.2 | NP_112353.1 | 2 | 304 | ||
XM_006238531.2 | XP_006238593.1 | 2 | 350 | |||
GenBank mRNA | BC060562.1 | - | 2 | 222 | 6918181 | |
Y14019.1 | - | 1 | 192 |
Gene Symbol: Rab3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S68807.1 | - | 147 |