Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031104.1 | NP_112366.1 | 3 | 105 | ||
GenBank mRNA | BC058466.1 | - | 2 | 121 | 6887477 | |
BC082750.1 | - | 2 | 96 | 7300771 | ||
FQ222147.1 | - | 2 | 114 | |||
FQ222660.1 | - | 2 | 114 | |||
FQ222798.1 | - | 2 | 115 | |||
FQ224693.1 | - | 2 | 113 | |||
FQ228373.1 | - | 2 | 111 | |||
X78444.1 | - | 3 | 105 |
Gene Symbol: LOC100360057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729392.4 | XP_002729438.2 | 1 | 118 |