Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031130.1 | NP_112392.1 | 1, 2 | 934 | ||
GenBank mRNA | U10995.1 | - | 1, 2 | 934 | ||
U15002.1 | - | 1, 2 | 120 |