Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031138.2 | NP_112400.1 | 1, 2 | 133 | ||
XM_006246343.2 | XP_006246405.1 | 2, 3 | 158 | |||
GenBank mRNA | BC070946.1 | - | 1, 2 | 126 | 7103580 | |
DN931313.1 | - | 1, 2 | 163 | |||
FQ222428.1 | - | 1, 2 | 150 | |||
FQ229894.1 | - | 1, 2 | 144 | |||
M62388.1 | - | 1, 2 | 75 |
Gene Symbol: LOC103694902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773893.1 | XP_008772115.1 | 2, 3 | 157 |