Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277211.1 | NP_001264140.1 | 7 | 736 | ||
NM_001277212.1 | NP_001264141.1 | 7 | 736 | |||
NM_001277213.1 | NP_001264142.1 | 7 | 736 | |||
NM_031352.2 | NP_112642.1 | 7 | 736 | |||
GenBank mRNA | AB009346.1 | - | 7 | 707 | ||
AB038364.1 | - | 7 | 665 | |||
AB038365.1 | - | 7 | 707 | |||
AB039818.1 | - | 7 | 673 | |||
AB039819.1 | - | 7 | 707 | |||
BC072483.1 | - | 7 | 713 | 7104722 | ||
FQ228854.1 | - | 7 | 736 |