Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031793.1 | NP_113981.1 | 15 | 1768 | ||
XM_006234324.2 | XP_006234386.1 | 15 | 2144 | |||
XM_006234325.2 | XP_006234387.1 | 15 | 1965 | |||
XM_006234326.2 | XP_006234388.1 | 15 | 2088 | |||
XM_006234327.2 | XP_006234389.1 | 14 | 2040 | |||
XM_006234328.2 | XP_006234390.1 | 15 | 2035 | |||
XM_006234330.2 | XP_006234392.1 | 14 | 1861 | |||
XM_006234331.2 | XP_006234393.1 | 14 | 1987 | |||
XM_006234333.2 | XP_006234395.1 | 14 | 2001 | |||
XM_006234334.2 | XP_006234396.1 | 14 | 1822 | |||
XM_006234335.2 | XP_006234397.1 | 14 | 1948 | |||
XM_008761930.1 | XP_008760152.1 | 14 | 2064 | |||
XM_008761931.1 | XP_008760153.1 | 14 | 2025 | |||
GenBank mRNA | AF043642.1 | - | 15 | 1768 | ||
BC085723.1 | - | 14 | 1814 | 7123868 |