Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031793.1 | NP_113981.1 | 11 | 962 | ||
XM_006234324.2 | XP_006234386.1 | 11 | 1338 | |||
XM_006234325.2 | XP_006234387.1 | 11 | 1159 | |||
XM_006234326.2 | XP_006234388.1 | 11 | 1282 | |||
XM_006234327.2 | XP_006234389.1 | 10 | 1234 | |||
XM_006234328.2 | XP_006234390.1 | 11 | 1229 | |||
XM_006234330.2 | XP_006234392.1 | 10 | 1055 | |||
XM_006234331.2 | XP_006234393.1 | 10 | 1181 | |||
XM_006234333.2 | XP_006234395.1 | 10 | 1195 | |||
XM_006234334.2 | XP_006234396.1 | 10 | 1016 | |||
XM_006234335.2 | XP_006234397.1 | 10 | 1142 | |||
XM_008761930.1 | XP_008760152.1 | 10 | 1258 | |||
XM_008761931.1 | XP_008760153.1 | 10 | 1219 | |||
GenBank mRNA | AF043642.1 | - | 11 | 962 | ||
BC085723.1 | - | 10 | 1008 | 7123868 | ||
FQ225838.1 | - | 6 | 477 | |||
FQ230740.1 | - | 6 | 477 | |||
FQ230786.1 | - | 6 | 477 | |||
FQ232477.1 | - | 6 | 478 |