Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032062.2 | NP_114451.2 | 88 | |||
GenBank mRNA | AF230644.1 | - | 88 | |||
AF232668.1 | - | 58 | ||||
AF232669.1 | - | 58 | ||||
AY621095.1 | - | 58 | ||||
U88156.1 | - | 441 | ||||
U88157.1 | - | 58 |