Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032069.1 | NP_114458.1 | 13 | 1893 | ||
GenBank mRNA | AY437398.1 | - | 880 | |||
DQ227255.1 | - | 4 | 533 | |||
DQ227256.1 | - | 1763 | ||||
U88572.1 | - | 13 | 1893 |