Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053399.1 | NP_445851.1 | 2 | 524 | ||
XM_006244347.2 | XP_006244409.1 | 3 | 933 | |||
GenBank mRNA | AB032562.1 | - | 1 | 54 | ||
AY190603.1 | - | 2 | 524 |