Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053419.1 | NP_445871.1 | 1855 | |||
GenBank mRNA | AF169549.1 | - | 826 | |||
AF242544.1 | - | 1513 | ||||
EF121977.1 | - | 1318 | ||||
EF121978.1 | - | 1318 | ||||
EF121979.1 | - | 1552 | ||||
EF121980.1 | - | 1552 | ||||
EF121981.1 | - | 1855 | ||||
EF121982.1 | - | 1318 |