Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053439.1 | NP_445891.1 | 6 | 660 | ||
XM_006249285.2 | XP_006249347.1 | 6 | 647 | |||
GenBank mRNA | AF306457.1 | - | 6 | 660 | ||
BC059123.1 | - | 6 | 675 | 6920235 | ||
FQ220083.1 | - | 6 | 643 | |||
FQ220326.1 | - | 6 | 643 | |||
FQ220799.1 | - | 6 | 688 | |||
FQ229342.1 | - | 6 | 612 | |||
FQ229588.1 | - | 6 | 666 | |||
FQ229761.1 | - | 6 | 651 | |||
FQ233248.1 | - | 6 | 614 |
Gene Symbol: Rasl2-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166675.1 | NP_001160147.1 | 1 | 746 | ||
GenBank mRNA | AF507943.1 | - | 1 | 625 |