Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053479.1 | NP_445931.1 | 28 | 3140 | ||
GenBank mRNA | AJ011605.1 | - | 28 | 3140 |