Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053489.2 | NP_445941.2 | 33 | 2785 | ||
GenBank mRNA | CA510744.1 | - | 3 | 140 |