Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109883.2 | NP_001103353.1 | 24 | 2522 | ||
GenBank mRNA | AF293383.1 | - | 23 | 2418 | ||
EV771702.1 | - | 7 | 575 |