Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053602.2 | NP_446054.1 | 2 | 283 | ||
XM_008768600.1 | XP_008766822.1 | 3 | 683 | |||
XM_008768601.1 | XP_008766823.1 | 3 | 407 | |||
XM_008768602.1 | XP_008766824.1 | 3 | 357 | |||
XM_008768603.1 | XP_008766825.1 | 2 | 257 | |||
GenBank mRNA | BC059121.1 | - | 2 | 191 | 6919556 | |
FQ212150.1 | - | 2 | 186 | |||
FQ214305.1 | - | 2 | 186 | |||
FQ216982.1 | - | 2 | 178 | |||
FQ217271.1 | - | 2 | 184 | |||
FQ217448.1 | - | 2 | 190 | |||
FQ224472.1 | - | 2 | 186 | |||
FQ228382.1 | - | 2 | 186 | |||
M73030.1 | - | 2 | 170 | |||
X54510.1 | - | 2 | 283 |