Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000389.4 | NP_000380.1 | 2 | 282 | ||
NM_001220777.1 | NP_001207706.1 | 2 | 227 | |||
NM_001220778.1 | NP_001207707.1 | 2 | 392 | |||
NM_001291549.1 | NP_001278478.1 | 3 | 434 | |||
NM_078467.2 | NP_510867.1 | 3 | 229 | |||
GenBank mRNA | AB209881.1 | - | 2 | 239 | ||
AB451290.1 | - | 1 | 157 | |||
AB451422.1 | - | 1 | 157 | |||
AK298901.1 | - | 2 | 291 | |||
AK309512.1 | - | 2, 3 | 251 | |||
BC000275.1 | - | 2 | 222 | 3355833 | ||
BC000312.2 | - | 2 | 376 | 2821049 | ||
BC001935.1 | - | 2 | 237 | 2823668 | ||
BC013967.2 | - | 2 | 240 | 2822909 | ||
BT006719.1 | - | 1 | 157 | |||
BU154329.1 | - | 3 | 250 | |||
BU193165.1 | - | 3 | 434 | |||
CR536533.1 | - | 1 | 157 | |||
DA552678.1 | - | 2 | 392 | |||
DA582789.1 | - | 2 | 283 | |||
DA699672.1 | - | 2 | 279 | |||
DA702185.1 | - | 2 | 227 | |||
DA838987.1 | - | 2 | 190 | |||
L25610.1 | - | 2 | 235 | |||
L26165.1 | - | 2 | 235 | |||
L47232.1 | - | 2 | 210 | |||
L47233.1 | - | 2 | 210 | |||
S67388.1 | - | 2 | 232 | |||
U03106.1 | - | 2 | 232 | |||
U09579.1 | - | 2 | 251 |