Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256799.2 | NP_001243728.1 | 6 | 724 | ||
NM_001289745.1 | NP_001276674.1 | 7 | 782 | |||
NM_001289746.1 | NP_001276675.1 | 6 | 676 | |||
NM_002046.5 | NP_002037.2 | 7 | 690 | |||
GenBank mRNA | AB062273.1 | - | 7 | 554 | ||
AF261085.1 | - | 7 | 605 | |||
AK026525.1 | - | 7 | 627 | |||
AK299448.1 | - | 7 | 575 | |||
AK308198.1 | - | 7 | 578 | |||
AY007133.1 | - | 7 | 554 | |||
AY633612.1 | - | 6 | 526 | |||
BC001601.1 | - | 7 | 556 | 3543589 | ||
BC004109.2 | - | 7 | 554 | 3678259 | ||
BC009081.1 | - | 7 | 563 | 3869809 | ||
BC013310.2 | - | 7 | 562 | 3538383 | ||
BC020308.1 | - | 6 | 540 | |||
BC023632.2 | - | 6 | 526 | 4842418 | ||
BC025925.1 | - | 7 | 559 | 2989348 | ||
BC026907.1 | - | 6 | 540 | 4454083 | ||
BC029340.1 | - | 7 | 548 | |||
BC029618.1 | - | 7 | 556 | 3876535 | ||
BC083511.1 | - | 7 | 554 | 3536969 | ||
BG724119.1 | - | 6 | 709 | |||
BI463134.1 | - | 6 | 707 | |||
BT006893.1 | - | 6 | 502 | |||
BU155402.1 | - | 7 | 727 | |||
CR407671.1 | - | 6 | 502 | |||
DQ403057.1 | - | 5 | 427 | |||
EF036498.1 | - | 3 | 240 | |||
EU668321.1 | - | 7 | 604 | |||
J02642.1 | - | 6 | 503 | |||
M17851.1 | - | 6 | 535 | |||
M33197.1 | - | 7 | 562 | |||
X01677.1 | - | 6 | 562 | |||
X53778.1 | - | 7 | 558 |