Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 2, 3 | 338 | ||
NM_018890.3 | NP_061485.1 | 2, 3 | 338 | |||
GenBank mRNA | AB209410.1 | - | 2, 3 | 285 | ||
AB451238.1 | - | 2, 3 | 97 | |||
AB451363.1 | - | 2, 3 | 97 | |||
AF136373.1 | - | 2, 3 | 97 | |||
AF498964.1 | - | 2, 3 | 97 | |||
AJ132694.1 | - | 2, 3 | 97 | |||
AK054993.1 | - | 2, 3 | 313 | |||
AK291806.1 | - | 2, 3 | 294 | |||
AK291815.1 | - | 2, 3 | 284 | |||
AY279384.1 | - | 2, 3 | 255 | |||
BC004247.1 | - | 2, 3 | 235 | 3610745 | ||
BC050687.1 | - | 2, 3 | 285 | 6149377 | ||
BC107748.1 | - | 2, 3 | 300 | 6699945 | ||
BG437715.1 | - | 2, 3 | 337 | |||
BQ689587.1 | - | 3, 4 | 354 | |||
BT007121.1 | - | 2, 3 | 97 | |||
M29870.1 | - | 2, 3 | 97 | |||
M31467.1 | - | 2, 3 | 97 |