Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164269.1 | NP_001157741.1 | 8, 9 | 916 | ||
NM_002875.4 | NP_002866.2 | 8, 9 | 1068 | |||
NM_133487.3 | NP_597994.3 | 8, 9 | 1071 | |||
XM_006720626.3 | XP_006720689.1 | 8, 9 | 1119 | |||
XM_011521857.2 | XP_011520159.1 | 8, 9 | 818 | |||
XM_011521858.2 | XP_011520160.1 | 8, 9 | 865 | |||
XM_011521859.2 | XP_011520161.1 | 8, 9 | 1002 | |||
XM_011521860.2 | XP_011520162.1 | 8, 9 | 913 | |||
XM_011521862.2 | XP_011520164.1 | 7, 8 | 918 | |||
GenBank mRNA | AK131299.1 | - | 8, 9 | 916 | ||
AK291969.1 | - | 8, 9 | 1026 | |||
AK313503.1 | - | 8, 9 | 1023 | |||
BC001459.2 | - | 6, 7 | 734 | 3139011 | ||
BT019705.1 | - | 7, 8 | 769 | |||
CR536559.1 | - | 7, 8 | 769 | |||
D13804.1 | - | 8, 9 | 980 | |||
D14134.1 | - | 8, 9 | 1001 |