Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 6 | 1151 | ||
NM_001143677.1 | NP_001137149.1 | 4 | 402 | |||
NM_001143678.1 | NP_001137150.1 | 4 | 581 | |||
NM_001291995.1 | NP_001278924.1 | 4 | 357 | |||
NM_005627.3 | NP_005618.2 | 4 | 357 | |||
XM_011536071.1 | XP_011534373.1 | 7 | 933 | |||
GenBank mRNA | AF153609.1 | - | 4 | 310 | ||
AK055077.1 | - | 6 | 1151 | |||
AK098509.1 | - | 4 | 325 | |||
AK293383.1 | - | 4 | 391 | |||
AK295422.1 | - | 4 | 325 | |||
AK298688.1 | - | 4 | 640 | |||
BC001263.1 | - | 4 | 310 | 3459056 | ||
BX649005.1 | - | 2 | 1196 | |||
DA309338.1 | - | 4 | 435 | |||
EU518415.1 | - | 6 | 553 | |||
FM205707.1 | - | 4 | 345 | |||
FM205708.1 | - | 4 | 380 | |||
FM205709.1 | - | 6 | 1000 | |||
FM205710.1 | - | 3 | 469 | |||
Y10032.1 | - | 4 | 310 |