Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 5 | 1089 | ||
NM_001143677.1 | NP_001137149.1 | 3 | 340 | |||
NM_001143678.1 | NP_001137150.1 | 3 | 519 | |||
NM_001291995.1 | NP_001278924.1 | 3 | 295 | |||
NM_005627.3 | NP_005618.2 | 3 | 295 | |||
XM_011536071.1 | XP_011534373.1 | 6 | 871 | |||
GenBank mRNA | AF153609.1 | - | 3 | 248 | ||
AK055077.1 | - | 5 | 1089 | |||
AK098509.1 | - | 3 | 263 | |||
AK293383.1 | - | 3 | 329 | |||
AK295422.1 | - | 3 | 263 | |||
AK298688.1 | - | 3 | 578 | |||
BC001263.1 | - | 3 | 248 | 3459056 | ||
BX649005.1 | - | 2 | 696 | |||
DA309338.1 | - | 3 | 373 | |||
EU518415.1 | - | 5 | 491 | |||
FM205707.1 | - | 3 | 283 | |||
FM205708.1 | - | 3 | 318 | |||
FM205709.1 | - | 5 | 938 | |||
FM205710.1 | - | 2 | 407 | |||
Y10032.1 | - | 3 | 248 |