Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243925.1 | NP_001230854.1 | 6 | 686 | ||
NM_001243926.1 | NP_001230855.1 | 8 | 1165 | |||
NM_004635.4 | NP_004626.1 | 6 | 671 | |||
GenBank mRNA | AB451303.1 | - | 5 | 523 | ||
AB451434.1 | - | 5 | 523 | |||
AK290969.1 | - | 6 | 642 | |||
AK313565.1 | - | 5 | 588 | |||
BC001662.2 | - | 6 | 632 | 2988489 | ||
BC007591.1 | - | 6 | 608 | 3345889 | ||
BC010407.2 | - | 6 | 630 | 3833667 | ||
BC068497.1 | - | 8 | 1119 | |||
U09578.1 | - | 6 | 614 | |||
U43784.1 | - | 5 | 545 |