Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243925.1 | NP_001230854.1 | 9 | 1048 | ||
NM_001243926.1 | NP_001230855.1 | 11 | 1527 | |||
NM_004635.4 | NP_004626.1 | 9 | 1033 | |||
GenBank mRNA | AB451303.1 | - | 8 | 885 | ||
AB451434.1 | - | 8 | 885 | |||
AK290969.1 | - | 9 | 1004 | |||
AK313565.1 | - | 8 | 950 | |||
BC001662.2 | - | 9 | 994 | 2988489 | ||
BC007591.1 | - | 9 | 970 | 3345889 | ||
BC010407.2 | - | 9 | 992 | 3833667 | ||
BC068497.1 | - | 11 | 1481 | |||
U09578.1 | - | 9 | 976 | |||
U43784.1 | - | 8 | 907 |