Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017572.3 | NP_060042.2 | 11 | 1100 | ||
NM_199054.2 | NP_951009.1 | 11 | 1100 | |||
GenBank mRNA | AB209074.1 | - | 3 | 271 | ||
AF125532.1 | - | 10 | 877 | |||
AF237775.1 | - | 10 | 891 | |||
AF237776.1 | - | 10 | 891 | |||
AK001730.1 | - | 1 | 151 | |||
AK293742.1 | - | 9 | 759 | |||
AK303393.1 | - | 8 | 770 | |||
AL137565.1 | - | 7 | 562 | |||
AL513974.3 | - | 1 | 211 | |||
BC018345.1 | - | 3 | 269 | |||
BC073140.1 | - | 11 | 1080 | 6527053 | ||
BM977403.1 | - | 4 | 673 |