Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256295.1 | NP_001243224.1 | 9 | 1423 | ||
NM_005239.5 | NP_005230.1 | 8 | 1253 | |||
XM_005260935.1 | XP_005260992.1 | 8 | 1326 | |||
XM_017028290.1 | XP_016883779.1 | 8 | 3923 | |||
GenBank mRNA | AK096841.1 | - | 9 | 1423 | ||
AK315563.1 | - | 8 | 1156 | |||
BC017040.1 | - | 8 | 1125 | 3852274 | ||
BC042954.2 | - | 8 | 1155 | 5276704 | ||
BT006838.1 | - | 7 | 963 | |||
J04102.1 | - | 8 | 1254 |