Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006609.4 | NP_006600.3 | 5 | 419 | ||
GenBank mRNA | AB208963.1 | - | 4 | 325 | ||
AF111105.1 | - | 5 | 420 | |||
AF239798.1 | - | 4 | 350 | |||
AK315007.1 | - | 6 | 603 | |||
BC136293.1 | - | 5 | 388 | 9020280 |