Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP4K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270425.1 | NP_001257354.1 | 28 | 2427 | ||
NM_003618.3 | NP_003609.2 | 29 | 2490 | |||
GenBank mRNA | AK291467.1 | - | 29 | 2456 | ||
AK299889.1 | - | 21 | 1519 | |||
AK314487.1 | - | 28 | 2264 | |||
AK316144.1 | - | 2 | 70 | |||
BC071579.1 | - | 28 | 2419 | 5296205 |