Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 8 | 917 | ||
NM_001300802.1 | NP_001287731.1 | 8 | 917 | |||
NM_001318361.1 | NP_001305290.1 | 8 | 871 | |||
NM_003942.2 | NP_003933.1 | 8 | 917 | |||
XM_005274380.1 | XP_005274437.1 | 8 | 897 | |||
XM_017018527.1 | XP_016874016.1 | 4 | 317 | |||
GenBank mRNA | AF074715.1 | - | 7 | 786 | ||
AJ010119.1 | - | 8 | 899 | |||
AK223561.1 | - | 8 | 922 | |||
AK292163.1 | - | 8 | 898 | |||
AK298507.1 | - | 8 | 842 | |||
AK313567.1 | - | 8 | 897 | |||
BC020451.1 | - | 5 | 472 | |||
BC028079.1 | - | 8 | 881 | |||
BC047896.1 | - | 8 | 896 | 5763859 |