Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135044.1 | NP_001128516.1 | 2, 3 | 387 | ||
NM_001308244.1 | NP_001295173.1 | 1, 2 | 163 | |||
NM_002752.4 | NP_002743.3 | 2, 3 | 387 | |||
NM_139068.2 | NP_620707.1 | 2, 3 | 387 | |||
NM_139069.2 | NP_620708.1 | 2, 3 | 387 | |||
NM_139070.2 | NP_620709.1 | 2, 3 | 387 | |||
XM_006714891.2 | XP_006714954.1 | 3, 4 | 445 | |||
XM_017009638.1 | XP_016865127.1 | 3, 4 | 356 | |||
XM_017009641.1 | XP_016865130.1 | 3, 4 | 454 | |||
XM_017009642.1 | XP_016865131.1 | 2, 3 | 283 | |||
XM_017009643.1 | XP_016865132.1 | 2, 3 | 283 | |||
GenBank mRNA | AB451302.1 | - | 1, 2 | 116 | ||
AB451355.1 | - | 1, 2 | 116 | |||
AB451433.1 | - | 1, 2 | 116 | |||
AK289638.1 | - | 2, 3 | 283 | |||
BC032539.1 | - | 1, 2 | 177 | 5528624 | ||
CR536580.1 | - | 1, 2 | 116 | |||
DB451562.1 | - | 2, 3 | 413 | |||
EU927388.1 | - | 1, 2 | 136 | |||
HM036706.1 | - | 1, 2 | 136 | |||
HM036707.1 | - | 1, 2 | 136 | |||
HM036708.1 | - | 1, 2 | 136 | |||
L31951.1 | - | 1, 2 | 174 | |||
U09759.1 | - | 1, 2 | 267 | |||
U34821.1 | - | 1, 2 | 160 | |||
U35002.1 | - | 1, 2 | 138 | |||
U35003.1 | - | 1, 2 | 150 |